Squibb Dropping Funnels | Avantor
Squibb Dropping Funnels
Socket Size 24/29, 500 ml
Catalog # 470314-008
Supplier:   Wards
Fournisseur #:  CH0476C
Squibb Dropping Funnels
Catalog # 470314-008
Supplier:  Wards
Squibb Dropping Funnels
Squibb Dropping Funnels
Socket Size 24/29, 500 ml
Price
54,95 $
Log in pour la disponibilité
Each
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Spécifications
  • Capacité:
    500 ml
  • Désignation:
    Socket Size 24/29