Squibb Dropping Funnels
Socket Size 19/26, 250 ml
Catalog # 470314-006
Supplier: Wards
Fournisseur #: CH0476B
Jump to:
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Spécifications
- Capacité:250 ml
- Désignation:Socket Size 19/26