Squibb Dropping Funnels | Avantor
Squibb Dropping Funnels
Socket Size 19/26, 50 ml
Catalog # 470314-004
Supplier:   Wards
Fournisseur #:  CH0476A50
Squibb Dropping Funnels
Catalog # 470314-004
Supplier:  Wards
Squibb Dropping Funnels
Squibb Dropping Funnels
Socket Size 19/26, 50 ml
Price
$32.40
Log in pour la disponibilité
Each
La quantité minimale de commande est de 2
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Spécifications
  • Capacité:
    50 ml
  • Désignation:
    Socket Size 19/26