Squibb Dropping Funnels | Avantor
Squibb Dropping Funnels
Socket Size 19/26, 100 ml
Catalog # 470314-002
Supplier:   Wards
Fournisseur #:  CH0476A
Squibb Dropping Funnels
Catalog # 470314-002
Supplier:  Wards
Squibb Dropping Funnels
Squibb Dropping Funnels
Socket Size 19/26, 100 ml
Price
$41.65
Log in pour la disponibilité
Each
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Spécifications
  • Capacité:
    100 ml
  • Désignation:
    Socket Size 19/26