Squibb Dropping Funnels
Socket Size 19/26, 100 ml
Catalog # 470314-002
Supplier: Wards
Fournisseur #: CH0476A
Jump to:
Product Details & Documents
Made of borosilicate glass, with interchangeable plastic stopper and glass stopcock.
Spécifications
- Capacité:100 ml
- Désignation:Socket Size 19/26