Buchner Filtering Flasks
Socket Size 24/29,, 1000 ml, —
Catalog # 470313-988
Supplier: Wards
Fournisseur #: CH0422E
Jump to:
Product Details & Documents
Buchner filtering flasks are made of high quality, heavy duty borosilicate glass.
Ordering information: Stopper not included.
Spécifications
- Capacité:1000 ml
- Désignation:Socket Size 24/29,
- Diameter ×H:—