Polypropylene Buchner Funnels | Avantor
Polypropylene Buchner Funnels
70 mm
Catalog # 470313-476
Supplier:   GSC International
Fournisseur #:  BFP70
Polypropylene Buchner Funnels
Catalog # 470313-476
Supplier:  GSC International
Polypropylene Buchner Funnels
Polypropylene Buchner Funnels
70 mm
Price
$12.80
Log in pour la disponibilité
Each Pack of 10
Product Details & Documents
The polypropylene Buchner funnels are two-piece, light-weight, and easy-to-clean.
They are fully autoclavable. The top portion snaps to the bottom forming a vacuum tight seal.
Spécifications
  • Top Diameter:
    70 mm