Polypropylene Buchner Funnels
100 mm
Catalog # 470313-386
Supplier: GSC International
Fournisseur #: BFP100
Jump to:
Product Details & Documents
The polypropylene Buchner funnels are two-piece, light-weight, and easy-to-clean.
They are fully autoclavable. The top portion snaps to the bottom forming a vacuum tight seal.
Spécifications
- Top Diameter:100 mm