MAT COMPLETE BUBBLE ESD EFB PU 4INX7FT
Catalog # 76315-216
Supplier: Ergomat
Fournisseur #: EFB0407
Jump to:
Product Details & Documents
Spécifications
- Powdered/Powder-free:Powder-free
- Cat. No.:77200-738
- Cleanroom class:100/ISO 5
- Disposable / reusable:Disposable
- Ambidextrous/Hand-specific:Ambidextrous
- Latex/Latex-free:Latex-free
- Anti-static:+
- Material:Nitrile
- Sterility:Non sterile