Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Serological Pipettes, Class A

1 ml, ±0.007 ml, Blue
Catalog # 470312-924
Supplier:   Wards
 :  CH0573A
Serological Pipettes, Class A
Catalog # 470312-924
Supplier:  Wards
Serological Pipettes, Class A
Serological Pipettes, Class A

1 ml, ±0.007 ml, Blue

Price
13,65 $
Log in pour la disponibilité
Each
Product Details & Documents

Perfect pipette for everyday laboratory use.

  • Reusable
  • Color-coded for easy sorting
  • Accuracy as per Class A and DIN ISO 835 standard

These class A serological pipettes are calibrated for delivery to jet. The last drop is expelled by blowing. Blue graduations and color coding make these pipettes very easy to use.

Spécifications
  • Capacité:
    1 ml
  • Color Code:
    Blue
  • Tolerance:
    ±0.007 ml