ESP™ Pipet Tip Reloading Systems, Neptune™
20 - 300 µl tips, bulk
  76311-244
   Waste Reducing|Sustainable Packaging
 :   Ohaus
 :  63300802
ESP™ Pipet Tip Reloading Systems, Neptune™
  76311-244
 :  Ohaus
   Waste Reducing|Sustainable Packaging
ESP™ Pipet Tip Reloading Systems, Neptune™
ESP™ Pipet Tip Reloading Systems, Neptune™
20 - 300 µl tips, bulk
$28.73
Pack of 10 Case of 50
Revolutionary ESP reload system offers the environmentally friendly reuse of existing trays.
  • Autoclavable
  • Good sustainability
  • Superior stability, firm placement
  • Maximum space saving design
Eco-friendly empty pipette tips trays are manufactured from recycled polypropylene. When combined with ESP™ reload systems, these tip trays reduce plastic waste by up to 90%.
Unique transfer system minimizes plastic waste by as much as 90%. Reload 10 trays in 90 seconds. Integrated transfer card prevents contamination by minimizing handling. Universal pipette tips are made from virgin polypropylene. The thin wall design provides an optimal seal. Tips featuring the S³ polymer deliver to within 0.1% of indicated volume, ensuring a more accurate application.
 : Certified free of RNase, DNase, pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
  • For Use With:
    20 - 300 µl tips, bulk
  • Cat. No.:
    76311-244
  • Environmental certification:
    FSC-STD-40-004 V3-1 and FSC-C154485; Empty rack intended for Reload and Bulk Refills
  • Environmentally Preferable:
    Waste Reducing|Sustainable Packaging