0,00 $
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
T3-301 LOW-HIGH TEMP WH 3 X 100
Catalog # MSPP-T3-301
Supplier:
GRAPHIC PRODUCTS INC MS
Expand image
T3-301 LOW-HIGH TEMP WH 3 X 100
Catalog # MSPP-T3-301
Supplier:
GRAPHIC PRODUCTS INC MS
Supplier Number:
T3-301
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International