Objectives for REBEL Microscopes, ECHO
100× Plan Apochromat Oil, CSC 0.17, 0.13 mm, N.A. 1.35
Catalog # 76307-590
Supplier: DISCOVER ECHO INC.
Supplier #: RMAG9112
Jump to:
Product Details & Documents
Spécifications
- Désignation:100× Plan Apochromat Oil, CSC 0.17
- Aperture:N.A. 1.35
- Working Distance:0.13 mm