Objectives for REBEL Microscopes, ECHO
40× Plan Apochromat with Correction Collar, CSC 0.17, 0.13 - 0.16 mm, N.A. 0.95
Catalog # 76307-586
Supplier: DISCOVER ECHO INC.
Supplier #: RMAG9404
Jump to:
Product Details & Documents
Specifications
- Désignation:40× Plan Apochromat with Correction Collar, CSC 0.17
- Aperture:N.A. 0.95
- Working Distance:0.13 - 0.16 mm
- Cat. No.:76307-586