Objectives for REBEL Microscopes, ECHO
40× Plan Apochromat, CSC 0.17, 0.15 mm, N.A. 0.95
Catalog # 76307-584
Supplier: DISCOVER ECHO INC.
: RMAG9402
Jump to:
Product Details & Documents
Specifications
- Désignation:40× Plan Apochromat, CSC 0.17
- Aperture:N.A. 0.95
- Working Distance:0.15 mm