Objectives for REBEL Microscopes, ECHO
20× Plan Apochromat, CSC 0.17, 0.6 mm, N.A. 0.75
Catalog # 76307-582
Supplier: DISCOVER ECHO INC.
Fournisseur #: RMAG9202
Jump to:
Product Details & Documents
Spécifications
- Désignation:20× Plan Apochromat, CSC 0.17
- Aperture:N.A. 0.75
- Working Distance:0.6 mm