Objectives for REBEL Microscopes, ECHO
4× Plan Apochromat, CSC 0.17, 16.6 mm, N.A. 0.13
  76307-578
 :  RMAG9042
Objectives for REBEL Microscopes, ECHO
  76307-578
Objectives for REBEL Microscopes, ECHO
4× Plan Apochromat, CSC 0.17, 16.6 mm, N.A. 0.13
Each
  • Désignation:
    4× Plan Apochromat, CSC 0.17
  • Aperture:
    N.A. 0.13
  • Working Distance:
    16.6 mm