Objectives for REBEL Microscopes, ECHO
2× Plan Apochromat, CSC 0.17, 6.2 mm, N.A. 0.08
Catalog # 76307-576
Supplier: DISCOVER ECHO INC.
Fournisseur #: RMAG9022
Jump to:
Product Details & Documents
Spécifications
- Désignation:2× Plan Apochromat, CSC 0.17
- Aperture:N.A. 0.08
- Working Distance:6.2 mm