Objectives for REBEL Microscopes, ECHO
100× Plan Achromat Oil Phase, CSC 0.17, 0.2 mm, N.A. 1.25
76307-570
- Désignation:100× Plan Achromat Oil Phase, CSC 0.17
- Aperture:N.A. 1.25
- Working Distance:0.2 mm