Objectives for REBEL Microscopes, ECHO
4× Plan Achromat Phase, LWD and CSC 0.17, 17 mm, N.A. 0.13
Catalog # 76307-562
Supplier: DISCOVER ECHO INC.
: RMAG0043
Jump to:
Product Details & Documents
Spécifications
- Désignation:4× Plan Achromat Phase, LWD and CSC 0.17
- Aperture:N.A. 0.13
- Working Distance:17 mm