Objectives for REBEL Microscopes, ECHO
100× Plan Achromat Oil, CSC 0.17, 0.2 mm, N.A. 1.25
76307-560
: RMAG1002
- Désignation:100× Plan Achromat Oil, CSC 0.17
- Aperture:N.A. 1.25
- Working Distance:0.2 mm