Objectives for REBEL Microscopes, ECHO
20× Plan Achromat, CSC 0.17, 1.6 mm, N.A. 0.40
Catalog # 76307-556
Supplier: DISCOVER ECHO INC.
: RMAG0202
Jump to:
Product Details & Documents
Spécifications
- Désignation:20× Plan Achromat, CSC 0.17
- Aperture:N.A. 0.40
- Working Distance:1.6 mm