Objectives for REBEL Microscopes, ECHO
10× Plan Achromat, CSC 0.17, 12 mm, N.A. 0.25
Catalog # 76307-554
Supplier: DISCOVER ECHO INC.
Fournisseur #: RMAG0102
Jump to:
Product Details & Documents
Spécifications
- Désignation:10× Plan Achromat, CSC 0.17
- Aperture:N.A. 0.25
- Working Distance:12 mm