Objectives for REBEL Microscopes, ECHO
Objectives for REBEL Microscopes, ECHO
17 mm, N.A. 0.13
Catalog # 76307-552
Objectives for REBEL Microscopes, ECHO
Catalog # 76307-552
Supplier:  
Objectives for REBEL Microscopes, ECHO
17 mm, N.A. 0.13
Discontinued
Product Details & Documents
Specifications
  • Aperture:
    N.A. 0.13
  • Working Distance:
    17 mm