Objectives for REBEL Microscopes, ECHO
2× Plan Achromat, CSC 0.17, 5 mm, N.A. 0.06
Catalog # 76307-550
Supplier: DISCOVER ECHO INC.
Supplier #: RMAG0022
Jump to:
Product Details & Documents
Specifications
- Désignation:2× Plan Achromat, CSC 0.17
- Aperture:N.A. 0.06
- Working Distance:5 mm