Laboratory Aprons
Regular, 36×27"
Catalog # 470311-690
Supplier: United Scientific Supplies
Fournisseur #: LBAP01
Jump to:
Product Details & Documents
Protective laboratory apron for multipurpose use.
- Flexible
- Chemical resistant
- Apron includes cloth ties at the neck and waist
These aprons are made of a PVC-coated fabric which stays flexible. There is reinforcement at the points of strain.
Spécifications
- Coupe du tenue:Regular
- L×l:36×27"
- Cat. No.:470311-690
- Garment application:Chemical protection|General Purpose|Laboratory
- Gender:Unisex
- Fabric:PVC-coated fabric