Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Trypsinizing Flasks, Bellco Glass

Trypsinizing flask, 250 ml, 80×140 mm
  76305-354
 :  1992-00250
Trypsinizing Flasks, Bellco Glass
  76305-354
Trypsinizing Flasks, Bellco Glass

Trypsinizing flask, 250 ml, 80×140 mm

 
6 703,10 $
   
Case of 3 (250ml)
 

Manufactured from Type 1 Class A Borosilicate Glass.

  • Designed with deep side baffles for optimal agitation
  • Large bottom surface for easy access with Bellco large orifice pipette
 
  • Désignation:
    Trypsinizing flask, 250 ml
  • O.D.×H:
    80×140 mm