Stirring Bars, Bellco
50.8 mm, 9.5 mm, White
Catalog # 76305-346
Marketsource
Supplier: Bellco Glass
Fournisseur #: 1975-00200
Jump to:
Product Details & Documents
Molded, seamless PTFE construction is ideal for procedures using all Bellco magnetic stirrers. Excellent for mixing media and buffer solutions.
Spécifications
- Ø:9.5 mm
- Color:White
- Longueur:50.8 mm



