Stirring Bars, Bellco
41.3 mm, 9.5 mm, White
Catalog # 76305-344
Supplier: Bellco Glass
Supplier #: 1975-00158
Jump to:
Product Details & Documents
Molded, seamless PTFE construction is ideal for procedures using all Bellco magnetic stirrers. Excellent for mixing media and buffer solutions.
Spécifications
- Ø:9.5 mm
- Color:White
- Longueur:41.3 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



