Ward's TracRack® | Avantor
Ward's TracRack®
0.5/0.2 ml
Catalog # 470311-182
Ward's TracRack®
Catalog # 470311-182
Supplier:  
Ward's TracRack®
Ward's TracRack®
0.5/0.2 ml
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
FREEZER RACK PP 96 PCR TUBE EA Catalog #470347-990
Le produit 470347-990 est un remplacement de 470311-182 mais varie selon la marque, l'emballage ou la quantité.
Product Details & Documents
Tube storage and a tube rack in one.
  • Helps reduce potential cross contamination
  • Saves valuable working space and allows more room for sample manipulation
  • Stops opened microfuge lids from interfering with other wells
  • Alphanumeric well identification
  • Tube segments sold with the box and lid
Removing and working with samples from cold temperature storage has never been easier than with the Ward's TracRack®. This rack and storage system eliminates the hassle of taking an entire rack of samples out of cold storage because the TracRack® enables lab professionals to remove one row of samples at a time.
Eight individual segments are nested in a 2" polypropylene film box that allows users to easily open, identify, and remove only the segments of samples needed. Once the segments are removed, simply replace the box lid and leave the remaining samples in storage. Each tube segment is made from durable polypropylene and allows for easy working while saving valuable counter space. Two versions are available.
Spécifications
  • For Tubes:
    0.5/0.2 ml