Polypropylene Stoppers | Avantor
Polypropylene Stoppers
24/29
Catalog # 470310-648
Supplier:   Wards
Fournisseur #:  CH0873E
Polypropylene Stoppers
Catalog # 470310-648
Supplier:  Wards
Polypropylene Stoppers
Polypropylene Stoppers
24/29
Price
8,35 $
Log in pour la disponibilité
Pack of 10
Product Details & Documents
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Spécifications
  • Stopper No.:
    24/29