Polypropylene Stoppers
19/26
Catalog # 470310-646
Supplier: Wards
Fournisseur #: CH0873D
Jump to:
Product Details & Documents
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Spécifications
- Stopper No.:19/26