Polypropylene Stoppers | Avantor
Polypropylene Stoppers
14/23
Catalog # 470310-644
Supplier:   Wards
Fournisseur #:  CH0873C
Polypropylene Stoppers
Catalog # 470310-644
Supplier:  Wards
Polypropylene Stoppers
Polypropylene Stoppers
14/23
Price
$7.05
Log in pour la disponibilité
Pack of 10
Product Details & Documents
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Spécifications
  • Stopper No.:
    14/23