Polypropylene Stoppers
10/19
Catalog # 470310-640
Supplier: Wards
Fournisseur #: CH0873A
Jump to:
Product Details & Documents
These lightweight and durable polypropylene stoppers are acid proof and autoclavable.
Spécifications
- Stopper No.:10/19