Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cryogenic Printable Labels, Brady

Cryogenic Printable Labels, Brady

Thermal transfer printer labels, Nylon Cloth, Silver, BBP®81, IP® series, PR Plus series, 1.75×1.875"
Catalog # 89209-495
Supplier:   Brady Worldwide
Supplier #:  104790
Cryogenic Printable Labels, Brady
Catalog # 89209-495
Supplier:  Brady Worldwide
Cryogenic Printable Labels, Brady
Cryogenic Printable Labels, Brady

Thermal transfer printer labels, Nylon Cloth, Silver, BBP®81, IP® series, PR Plus series, 1.75×1.875"

Price
$1,796.39
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each Roll of 1
Minimum order quantity is 1
Product Details & Documents

Cryogenic labels for tubes, vials and canes.

Chemical and stain resistant labels for slides and cassettes:
Temperatures from –80 to 121 °C (–112 to 250 °F).
B-449 Slide label with removability and write on capabilities.
B-481 Stainerbondz™ extreme chemical resistant slide label.
B-482 Tissue cassette label applied pre-process with the BSP®31 for extreme solvent/stain resistance.
B-488 Slide label with moderate chemical.
B-498 Tissue cassette label for re-identification of processed paraffin blocks.

General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more:
B-424 Economical paper label for general purpose.
B-427 Straw label for wrapping; working temperatures from –196 to 80 °C (–321 to 176 °F).
B-430 Petri dish and bag label; working temperatures from –80 to 121 °C (–112 to 250 °F).
B-499 For cryo boxes, freezer racks, petri dishes, bottles, flasks, microplates, and bags; working temperatures from –196 to 121 °C (–321 to 250 °F).

Spécifications
  • Pour l'utilisation avec:
    BBP®81, IP® series, PR Plus series
  • Color:
    Silver
  • Désignation:
    Thermal transfer printer labels
  • Dimensions:
    1.75×1.875"
  • Matière:
    Nylon Cloth