Objectives for REVOLVE Microscopes, Echo
60× U-Plan fluorite objective, 60×, 0.2 mm, 0.9 NA
Catalog # 76299-624
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0609
Jump to:
Product Details & Documents
Spécifications
- Grossissement:60×
- Désignation:60× U-Plan fluorite objective
- Aperture:0.9 NA
- Working Distance:0.2 mm