Objectives for REVOLVE Microscopes, Echo
4× POL U-Plan fluorite objective, 4×, 17 mm, 0.13 NA
  76299-622
 :  JMAG0046
Objectives for REVOLVE Microscopes, Echo
  76299-622
 :  
Objectives for REVOLVE Microscopes, Echo
4× POL U-Plan fluorite objective, 4×, 17 mm, 0.13 NA
  • Grossissement:
  • Désignation:
    4× POL U-Plan fluorite objective
  • Aperture:
    0.13 NA
  • Working Distance:
    17 mm