Objectives for REVOLVE Microscopes, Echo
10× C plan fluorite phase objective, 10×, 9.5 mm, 0.3 NA
Catalog # 76299-618
Marketsource
Supplier: DISCOVER ECHO INC.
Supplier #: JMAG0109
Objectives for REVOLVE Microscopes, Echo
Catalog # 76299-618
Supplier: DISCOVER ECHO INC.
Marketsource
Jump to:
Product Details & Documents
Spécifications
- Grossissement:10×
- Désignation:10× C plan fluorite phase objective
- Aperture:0.3 NA
- Working Distance:9.5 mm