Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Dissection Placemats | Avantor

Dissection Placemats

Dogfish Anatomy Placemat, Polyvinyl, 40 cm, 24.7 cm
Catalog # 470310-252
Supplier:   Ward's Science
Fournisseur #:  420.5045.1
Dissection Placemats
Catalog # 470310-252
Supplier:  Ward's Science
Dissection Placemats
Dissection Placemats

Dogfish Anatomy Placemat, Polyvinyl, 40 cm, 24.7 cm

Price
$21.35
Log in pour la disponibilité
Each
Product Details & Documents

Perfect Instructional Companion for Wet Lab Areas

  • Resistant to spills of any kind

Illustrations and descriptive text are printed right on the durable polyvinyl mat, so students can have specimen classification and information on the internal and external anatomy right on the lab bench with them while they are dissecting. Can be easily cleaned with a sponge or a paper towel, so students don’t have to worry about damaging a text book or other paper copies.

Spécifications
  • Largeur:
    24.7 cm
  • Désignation:
    Dogfish Anatomy Placemat
  • Longueur:
    40 cm
  • Finish:
    Polyvinyl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.