VWR® Microfiber String Mop | Avantor
VWR® Microfiber String Mop
Microfiber string mop, 12 oz.
Catalog # 76292-772
VWR® Microfiber String Mop
Catalog # 76292-772
Supplier:  
VWR® Microfiber String Mop
VWR® Microfiber String Mop
Microfiber string mop, 12 oz.
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VWR MOP MICROFIBER-STRING CS24 Catalog #89214-778
Le produit 89214-778 est similaire à 76292-772 mais le fabricant et les spécifications peuvent varier.
MOP MICROFIBER 1EA Catalog #89496-572
Le produit 89496-572 est similaire à 76292-772 mais le fabricant et les spécifications peuvent varier.
Product Details & Documents
This Microfiber String Mop is an absorbent and hard wearing mop for cleaning and liquid pick-up in controlled and critical environments.
  • Highly absorbent
  • Durable
The unique ribbing of the microfiber strands produces a mop that is both low linting and absorbent. The strands are comprised of split fiber polyester filaments, 30 times smaller than cotton fibers. These microfibers work to pick up tiny particles where cotton/rayon blends and polyesters may often roll over the dirt. The solid constructions of the mop, combined with the durable microfiber, allow the mop to be used with strong disinfectants, laundered and re-used.
Documents
Spécifications
  • Désignation:
    Microfiber string mop
  • Weight:
    12 oz.