Estwing Chisel Head Hammers
Chisel Hammer, 24 oz., 11.25"
Catalog # 470309-722
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
HAMMER-POL CHISEL HEAD 20 OZ Catalog #470309-720
Product 470309-720 is similar to 470309-722 but manufacturer and specifications may vary.
Product Details & Documents
Ideal for use with sedimentary rocks.
- Availabile in different head weights
Patented nylon end caps now makes these hammers the most durable on the market. The head and handles are forged in one piece and are fully polished. These tools have unsurpassed balance and are double tempered for added strength.
Spécifications
- Désignation:Chisel Hammer, 24 oz.
- Longueur:11.25"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



