Ao Absorbance Microplate Reader, Azure Biosystems
Ao 570 nm Filter
Catalog # 76292-010
 :  AC3107
Ao Absorbance Microplate Reader, Azure Biosystems
Catalog # 76292-010
Supplier:  
Ao Absorbance Microplate Reader, Azure Biosystems
Ao Absorbance Microplate Reader, Azure Biosystems
Ao 570 nm Filter
Discontinued
Product Details & Documents
96 well plate absorbance microplate reader.
  • Offers a simple user interface
  • Absorbance-based reading offers maximum sensitivity
  • Best read speeds for kinetic assays
  • High flexibility
  • Lower cost compared to fluorescent and luminescent-based readers
Its top-reading design is ideal for solution-based assays such as ELISAs, DNA and protein quantification, and enzyme kinetics.
Stand-alone, top absorbance reader. Large, easy-to-use 7" touch screen interface. Pre-set and custom protocols. Single and dual wavelength detection modes. 8 position filter wheel: 5 standard filters (405, 450, 492, 595, and 630) plus 3 options from 340 to 750 nm. Built-in shaker with speed selection. Motorized door for easy sample loading.
Spécifications
  • Désignation:
    Ao 570 nm Filter
Services
Informations sur le produit
  • Catalog #
    Description
    Availability
    Price
  • 76291-984
    Additional 1-Year Warranty, Plate Reader
    Log in pour la disponibilité
    Each - $1,305.21