Ao Absorbance Microplate Reader, Azure Biosystems
Ao 415 nm Filter
  76291-992
 :  AC3103
Ao Absorbance Microplate Reader, Azure Biosystems
  76291-992
Ao Absorbance Microplate Reader, Azure Biosystems
Ao Absorbance Microplate Reader, Azure Biosystems
Ao 415 nm Filter
$382.38
Each
96 well plate absorbance microplate reader.
  • Offers a simple user interface
  • Absorbance-based reading offers maximum sensitivity
  • Best read speeds for kinetic assays
  • High flexibility
  • Lower cost compared to fluorescent and luminescent-based readers
Its top-reading design is ideal for solution-based assays such as ELISAs, DNA and protein quantification, and enzyme kinetics.
Stand-alone, top absorbance reader. Large, easy-to-use 7" touch screen interface. Pre-set and custom protocols. Single and dual wavelength detection modes. 8 position filter wheel: 5 standard filters (405, 450, 492, 595, and 630) plus 3 options from 340 to 750 nm. Built-in shaker with speed selection. Motorized door for easy sample loading.
  • Désignation:
    Ao 415 nm Filter
Services
  •  
    Description
  • 76291-984
    Additional 1-Year Warranty, Plate Reader
    Each - $1,305.21