Ao Absorbance Microplate Reader, Azure Biosystems
Ao 415 nm Filter
76291-992
: AC3103
96 well plate absorbance microplate reader.
- Offers a simple user interface
- Absorbance-based reading offers maximum sensitivity
- Best read speeds for kinetic assays
- High flexibility
- Lower cost compared to fluorescent and luminescent-based readers
Its top-reading design is ideal for solution-based assays such as ELISAs, DNA and protein quantification, and enzyme kinetics.
Stand-alone, top absorbance reader. Large, easy-to-use 7" touch screen interface. Pre-set and custom protocols. Single and dual wavelength detection modes. 8 position filter wheel: 5 standard filters (405, 450, 492, 595, and 630) plus 3 options from 340 to 750 nm. Built-in shaker with speed selection. Motorized door for easy sample loading.
- Désignation:Ao 415 nm Filter