Botley™ The Coding Robot Classroom Set, Learning Resources | Avantor
Botley™ The Coding Robot Classroom Set, Learning Resources
Botley™ Single
Catalog # 470309-514
Supplier:   Learning Resources
Fournisseur #:  LER2936
Botley™ The Coding Robot Classroom Set, Learning Resources
Catalog # 470309-514
Supplier:  Learning Resources
Botley™ The Coding Robot Classroom Set, Learning Resources
Botley™ The Coding Robot Classroom Set, Learning Resources
Botley™ Single
Price
$116.40
Log in pour la disponibilité
Each
Product Details & Documents
Bring coding to the classroom!
  • 100% screen-free coding; no app, tablet, or phone required
  • 239-piece set has four Botley™ the Coding Robots
  • Discover advanced features including object detection, looping commands, and black-line following
  • Build critical thinking skills by helping Botley™ navigate obstacle courses or solve chain reaction puzzles
Botley™ the Coding Robot Classroom Set from Learning Resources contains everything teachers need to introduce early coding fundamentals to students as young as 5. Ready to use right out of the box, Botley™ teaches coding lessons that grow with students’ skills though 100% screen-free play (no app, tablet, or phone required). After they’ve mastered basic coding commands, students can use advanced features such as object detection, looping commands, and a black-line-following mode to help navigate through obstacle courses, solve chain-reaction puzzles, or travel around the classroom. In addition to building early coding skills, the device also helps students master logic, critical thinking, and other STEM skills.
Delivery information: The 239-piece Botley™ the Coding Robot Classroom Set comes with 2×77-piece Botley™ the Coding Robot Activity Sets, 2×additional 45-piece Botley™ the Coding Robots, and 1×41-piece Botley™ the Coding Robot Action Challenge Accessory Set.
Spécifications
  • Désignation:
    Botley™ Single