Eisco® Individual Bones | Avantor
Eisco® Individual Bones
Humerus, Left Side
Catalog # 470308-090
Supplier:   Wards
Fournisseur #:  AMCH1035AS
Eisco® Individual Bones
Catalog # 470308-090
Supplier:  Wards
Eisco® Individual Bones
Eisco® Individual Bones
Humerus, Left Side
Price
$30.50
Log in pour la disponibilité
Each
Product Details & Documents
For Individual Study.
  • Sold in left and right side variants
  • Great addition for your current skeleton
These individual bones are a great addition to you collection. They can be used as a match for your existing Eisco disarticulated skeleton, or to allow students to compare general skeletal structure. Additionally, these can be used as additional bones to simulate forensics activities, and skeletal reconstruction labs. Unless otherwise indicated, bones are not prepared in an articulated form, and have not been drilled.
Spécifications
  • Désignation:
    Humerus, Left Side