AccuScript High-Fidelity Reverse Transcriptase, Agilent Technologies
AccuScript high-fidelity 1st strand cDNA synthesis kit, The kit delivers the highest accuracy and offers you greater flexibility to use the cDNA in your choice of downstream applications such as PCR amplification or real-time PCR quantification, 50
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Fournisseur #: 200820
AccuScript High-Fidelity Reverse Transcriptase, Agilent Technologies
Catalog # CAST200820
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
AccuScript High-Fidelity Reverse Transcriptase delivers the highest RT accuracy currently available.
- MMLV-RT derivative combined with 3 to 5’ exonuclease reduces errors
- High yields of full length cDNA up to 20 kb;RT-PCR kit synthesises cDNA from 10 to 1000 ng of total RNA or 0.1 to 10 ng of poly(A)+ RNA
- Stringent quality control procedures ensure high yields of premium cDNA
AccuScript High-Fidelity Reverse Transcriptase (RT) delivers 3.7 times more accuracy than other commercially available RTs and 6.6 times more accuracy than AMV RT. The AccuScript PfuUltra II RT-PCR kit provides a complete system in applications that demand high fidelity, specificity, and throughput.
Caution: For research use only. Not for use in diagnostic procedures.
Spécifications
Spécifications
- Désignation:AccuScript high-fidelity 1st strand cDNA synthesis kit
- No. of Reactions:50
- Details:The kit delivers the highest accuracy and offers you greater flexibility to use the cDNA in your choice of downstream applications such as PCR amplification or real-time PCR quantification
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



