pBlueScript II Vectors, Agilent Technologies | Avantor
pBlueScript II Vectors, Agilent Technologies
pBluescript II KS(–) Phagemid Kit, 1 Kit, f1 origin in (−) orientation, Kpn-Sac polylinker orientation. Contains: 20 ug pBluescript II KS(−) phagemid vector, host strain: XL1-Blue MRF'
Catalog # CA99900-360
pBlueScript II Vectors, Agilent Technologies
Catalog # CA99900-360
Supplier:  
CAS Number:  
pBlueScript II Vectors, Agilent Technologies
pBlueScript II Vectors, Agilent Technologies
pBluescript II KS(–) Phagemid Kit, 1 Kit, f1 origin in (−) orientation, Kpn-Sac polylinker orientation. Contains: 20 ug pBluescript II KS(−) phagemid vector, host strain: XL1-Blue MRF'
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
  • High copy number ColE1-based phagemid
  • Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
  • T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
Caution: For research use only. Not for use in diagnostic procedures.
Spécifications
Spécifications
  • Désignation:
    pBluescript II KS(–) Phagemid Kit
  • Taille:
    1 Kit
  • Details:
    f1 origin in (−) orientation, Kpn-Sac polylinker orientation. Contains: 20 ug pBluescript II KS(−) phagemid vector, host strain: XL1-Blue MRF'