Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
AffinityScript cDNA Synthesis Kits, Agilent Technologies | Avantor

AffinityScript cDNA Synthesis Kits, Agilent Technologies

AffinityScript one-step RT-PCR Kit, Reduced PCR cycling times and a one-tube, master mix format. Combined performance of Herculase II fusion DNA polymerase and the ArchaeMaxx PCR enhancing factor, delivering superior PCR yield and reliability. Robust performance for difficult and GC-rich targets. Optimized for target lengths up to 9.5 kb., 100
Catalog # CA97066-440
Fournisseur #:  600188
AffinityScript cDNA Synthesis Kits, Agilent Technologies
Catalog # CA97066-440
CAS Number:  
AffinityScript cDNA Synthesis Kits, Agilent Technologies
AffinityScript cDNA Synthesis Kits, Agilent Technologies

AffinityScript one-step RT-PCR Kit, Reduced PCR cycling times and a one-tube, master mix format. Combined performance of Herculase II fusion DNA polymerase and the ArchaeMaxx PCR enhancing factor, delivering superior PCR yield and reliability. Robust performance for difficult and GC-rich targets. Optimized for target lengths up to 9.5 kb., 100

CAS Number:  
Price
$661.11
Log in pour la disponibilité
Kit of 2
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Provides the highest efficiency of conversion of RNA to cDNA, fully optimized for qPCR applications.

  • Optimized for real-time quantitative PCR (qPCR) application;Produces cDNA in 15 min with linear detection from 3 pg to 3 µg total RNA
  • Creates up to 12 kb cDNA in 25 min or less
  • Available as a first-strand synthesis kit and a two-step QRT-PCR master mix format

The AffinityScript qPCR cDNA Synthesis Kit offers efficient conversion of RNA to cDNA. Fully optimized for real-time quantitative PCR (qPCR) applications, the kit utilizes our qPCR-grade AffinityScript Multiple Temperature Reverse Transcriptase. It includes a buffer system optimized for real-time qPCR.

The streamlined protocol produces cDNA in 15 min with linear detection from 3 pg to 3 µg of total RNA. The easy-to-use process generates up to 12 kb cDNA in 25 min or less. Available as a first-strand synthesis kit and a two-step QRT-PCR master mix format, using SYBR Green or probe-based detection.

The AffinityScript Multiple Temperature cDNA synthesis kit contains Agilent’s popular, highly thermostable, qPCR-grade AffinityScript Multiple Temperature Reverse Transcriptase for robust yields, from 37 to 60 °C, with nearly full activity between 45 to 55 °C. With the AffinityScript multiple temperature cDNA synthesis kit, Agilent offers enzymes with a broad thermal profile.

The Agilent AffinityScript Multiple Temperature Reverse Transcriptase (RT) system is optimized to produce high cDNA yields over the broadest temperature range. During analysis the multi-temp RT device also ensures high affinity to primer-template complexes. cDNA can be synthesized in amounts up to 20 kb.

The system’s multi-temperature capability allows you to change RT reaction temperature without having to change the reverse transcriptase, for a faster and more efficient analysis. In addition, when amplifying low RNA input amounts and resolving RNA secondary structures it delivers high cDNA yields, and can operate effectively at temperatures up to 55 °C.

Caution: For research use only. Not for use in diagnostic procedures.

Spécifications

Spécifications

  • Désignation:
    AffinityScript one-step RT-PCR Kit
  • No. of Reactions:
    100
  • Details:
    Reduced PCR cycling times and a one-tube, master mix format. Combined performance of Herculase II fusion DNA polymerase and the ArchaeMaxx PCR enhancing factor, delivering superior PCR yield and reliability. Robust performance for difficult and GC-rich targets. Optimized for target lengths up to 9.5 kb.

Spécifications