pBlueScript II Vectors, Agilent Technologies | Avantor
pBlueScript II Vectors, Agilent Technologies
pBluescript II XR Predigested Vector, 55 Reactions, XR vector is predigested with Xho I and EcoR I
Catalog # CA95040-826
pBlueScript II Vectors, Agilent Technologies
Catalog # CA95040-826
Supplier:  
CAS Number:  
pBlueScript II Vectors, Agilent Technologies
pBlueScript II Vectors, Agilent Technologies
pBluescript II XR Predigested Vector, 55 Reactions, XR vector is predigested with Xho I and EcoR I
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
The pBluescript II phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
  • High copy number ColE1-based phagemid
  • Large and versatile polylinker in two orientations, f1 origin available in either (+) or (–) orientation
  • T3 and T7 promoters for in vitro transcription of RNA
The pBluescript II phagemids have an extensive polylinker with 21 unique restriction enzyme recognition sites.
Caution: For research use only. Not for use in diagnostic procedures.
Spécifications
Spécifications
  • Désignation:
    pBluescript II XR Predigested Vector
  • Taille:
    55 Reactions
  • Details:
    XR vector is predigested with Xho I and EcoR I