Multiple Affinity Removal Starter Reagent Kit, Agilent Technologies
Spin Concentrators, to concentrate proteins via low-speed centrifugation through 5K MWCo filters into a collection tube, 4 ml
Catalog # CA76204-918
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Supplier #: 5185-5991
Multiple Affinity Removal Starter Reagent Kit, Agilent Technologies
Catalog # CA76204-918
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Starter Reagent Kit for Multiple Affinity Removal LC Columns.
Delivery information: Includes: 2 bottles Buffer A (1 l), 1 bottle Buffer B (1 l), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator
Specifications
Specifications
- Désignation:Spin Concentrators
- Taille:4 ml
- Details:to concentrate proteins via low-speed centrifugation through 5K MWCo filters into a collection tube



