Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Multiple Affinity Removal Column - Ms-3 (Mouse), 4.60 mm, 50 mm
Catalog # CA76204-858
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Fournisseur #: 5188-5217
Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Catalog # CA76204-858
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
The Multiple Affinity Removal LC Column – Mouse 3 is designed as a simple way to remove THREE interfering high-abundant proteins from mouse plasma samples.
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
: For research use only.
Spécifications
- Ø int.:4.60 mm
- Désignation:Multiple Affinity Removal Column - Ms-3 (Mouse)
- Longueur:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



